missing translation for 'onlineSavingsMsg'
Learn More

1,10-Phenanthroline, 99%, may contain up to 1.5% water

Catalog No. AAA1316306
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1316306 5 g
AAA1316314 25 g
AAA1316322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1316306 Supplier Thermo Scientific Chemicals Supplier No. A1316306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 66-71-7 | C12H8N2 | 180.21 g/mol

1,10-Phenanthroline, anhydrous is used as a chelating agent, used as bidendate nitrogen ligands in Heck type reactions as an alternative to phospine ligands. It is widely used as an indicator for alkyllithium reagents and an inhibitor of metallopeptidase enzymes. It is also used in the analysis formulations in end-user industries like food, beverage and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,10-Phenanthroline is used as a chelating agent, used as bidendate nitrogen ligands in Heck type reactions as an alternative to phospine ligands. It is widely used as an indicator for alkyllithium reagents and an inhibitor of metallopeptidase enzymes. It is also used in the analysis formulations in end-user industries like food, beverage and pharmaceuticals.

Solubility
Soluble in acetone and ethanol. Slightly soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong acids. Store in a tightly closed container. Store in a cool, dry, well-ventilated area.

Chemical Identifiers

CAS 66-71-7
Molecular Formula C12H8N2
Molecular Weight (g/mol) 180.21
MDL Number MFCD00011678
InChI Key DGEZNRSVGBDHLK-UHFFFAOYSA-N
Synonym o-phenanthroline, 4,5-diazaphenanthrene, orthophenanthroline, phenanthroline, 1,10-o-phenanthroline, phen, 2-phenanthroline, beta-phenanthroline, .beta.-phenanthroline
PubChem CID 1318
ChEBI CHEBI:44975
IUPAC Name 1,10-phenanthroline
SMILES C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1

Specifications

Melting Point 116°C to 119°C
Boiling Point >330°C
Odor Odorless
Quantity 5 g
UN Number UN2811
Beilstein 126461
Sensitivity Hygroscopic
Merck Index 14,7214
Solubility Information Soluble in acetone and ethanol. Slightly soluble in water.
Formula Weight 180.21
Percent Purity 99%
Chemical Name or Material 1,10-Phenanthroline
Show More Show Less
Hazard Category H301-H373
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 200-629-2
RTECSNumber SF8300000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only