missing translation for 'onlineSavingsMsg'
Learn More

1,10-Phenanthroline-5,6-dione 98.0+%, TCI America™

Catalog No. P19731G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P19731G 1 g
P19735G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P19731G Supplier TCI America Supplier No. P19731G

Chemical Identifiers

CAS 27318-90-7
Molecular Formula C12H6N2O2
Molecular Weight (g/mol) 210.19
MDL Number MFCD00014473
InChI Key KCALAFIVPCAXJI-UHFFFAOYSA-N
Synonym 1,10 phenanthroline-5,6-dione, 1 10-phenanthroline-5 6-dione 97, pyridino 3,2-h quinoline-5,6-dione, phen-5,6-dione, pubchem16755, acmc-209gwd, d0jh7c, phox31, ksc204s8l, 1,2-dione-based compound, 17
PubChem CID 72810
IUPAC Name 5,6-dihydro-1,10-phenanthroline-5,6-dione
SMILES O=C1C(=O)C2=CC=CN=C2C2=NC=CC=C12

Specifications

Melting Point 257°C
Color Yellow
Quantity 1 g
Formula Weight 210.19
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,10-Phenanthroline-5,6-dione