missing translation for 'onlineSavingsMsg'
Learn More

1,10-Phenanthroline-2,9-dicarboxylic acid hydrate, 97%

Catalog No. p-7024224
Change view
Click to view available options
Quantity:
0.25 g
1 g
Catalog No. Quantity
AAA1854801 0.25 g
AAA1854803 1 g
2 options

Catalog No. AAA1854801

Supplier: Thermo Scientific Chemicals A1854801

Only null left

CAS: 57709-61-2 | C14H8N2O4 | 268.23 g/mol

1,10-Phenanthroline-2,9-dicarboxylic acid hydrate is used to produce 1,10-phenanthroline-2,9-dicarbonyl chloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,10-Phenanthroline-2,9-dicarboxylic acid hydrate is used to produce 1,10-phenanthroline-2,9-dicarbonyl chloride.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents, bases and reducing agents.

Chemical Identifiers

CAS 57709-61-2
Molecular Formula C14H8N2O4
Molecular Weight (g/mol) 268.23
MDL Number MFCD00209670
InChI Key FXSVCROWUPWXBP-UHFFFAOYSA-N
Synonym 1,10-phenanthroline-2,9-dicarboxylicacid, 2,9-dicarboxy-1,10-phenanthroline, hydrate, acmc-20ap89, 1,10 phenanthroline-2,9-dicarboxylic acid, pyridino 3,2-h quinoline-2,9-dicarboxylic acid
PubChem CID 7567134
SMILES OC(=O)C1=NC2=C(C=CC3=C2N=C(C=C3)C(O)=O)C=C1

Specifications

Quantity 0.25 g
IUPAC Name 1,10-phenanthroline-2,9-dicarboxylic acid
Formula Weight 268.23 (Anhydrous)
Percent Purity 98%
Chemical Name or Material 1,10-Phenanthroline-2,9-dicarboxylic acid hydrate
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only