missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,10-Phenanthroline, 0.3% w/v Acidified, Reagents
1,10-Phenanthroline, 0.3% w/v Acidified, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc CP121800500A
Specifications
| 1,10-Phenanthroline, 0.3% w/v Acidified | |
| Colorless | |
| 100°C | |
| H2O | |
| 1.001 | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water | |
| 0.3% | |
| Liquid |
| 7732-18-5,5144-89-8,7647-01-0 | |
| 1.9 | |
| Poly Bottle | |
| 500 mL | |
| o-Phenanthroline | |
| O | |
| 18.02 | |
| Reagent |
Chemical Identifiers
| 7732-18-5 | |
| 18.02 | |
| o-Phenanthroline | |
| O |
| H2O | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water |