missing translation for 'onlineSavingsMsg'
Learn More

1,10-Decanediol, 97%

CAS: 112-47-0 | C10H22O2 | 174.284 g/mol

Supplier:  Thermo Scientific Chemicals A1392322

 View more versions of this product

Catalog No. AAA1392322


Only null left

Description

Description

1,10-Decanediol is an minor intermediate in the production of polyesters. It also used in essence, perfumes and in pharmaceuticals. The diacrylate is hydrophobic and used for flexible dental materials, radiation curable coatings, and for lenses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,10-Decanediol is an minor intermediate in the production of polyesters. It also used in essence, perfumes and in pharmaceuticals. The diacrylate is hydrophobic and used for flexible dental materials, radiation curable coatings, and for lenses.

Solubility
Soluble in alcohol. Insoluble in water, petroleum ether.

Notes
Store in a cool, dry, well-ventilated area. Incompatible with oxidizing agent.
Specifications

Specifications

1,10-Decanediol
71°C to 74°C
297°C
C10H22O2
100 g
14,2849
Soluble in alcohol. Insoluble in water,petroleum ether.
C(CCCCCO)CCCCO
174.284
174.28
112-47-0
1.08
152°C (306°F)
MFCD00004749
1698975
1,10-decanediol, decamethylene glycol, decamethylenediol, 1,10-dihydroxydecane, 1,10-decamethylenediol, 1,10-decamethylene diol, 1,6-bis 2-hydroxyethyl hexane, alpha,omega-decanediol, unii-5i577udk52, 1,10-ddecanediol
FOTKYAAJKYLFFN-UHFFFAOYSA-N
decane-1,10-diol
37153
97%

Chemical Identifiers

112-47-0
174.284
FOTKYAAJKYLFFN-UHFFFAOYSA-N
37153
C(CCCCCO)CCCCO
C10H22O2
MFCD00004749
1,10-decanediol, decamethylene glycol, decamethylenediol, 1,10-dihydroxydecane, 1,10-decamethylenediol, 1,10-decamethylene diol, 1,6-bis 2-hydroxyethyl hexane, alpha,omega-decanediol, unii-5i577udk52, 1,10-ddecanediol
decane-1,10-diol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 203-975-2

RTECSNumber : HD8433713

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only