missing translation for 'onlineSavingsMsg'
Learn More

1,10-Decanediol, 97%

Catalog No. p-7021543
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1392322 100 g
AAA1392336 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1392322 Supplier Thermo Scientific Chemicals Supplier No. A1392322

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 112-47-0 | C10H22O2 | 174.284 g/mol

1,10-Decanediol is an minor intermediate in the production of polyesters. It also used in essence, perfumes and in pharmaceuticals. The diacrylate is hydrophobic and used for flexible dental materials, radiation curable coatings, and for lenses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,10-Decanediol is an minor intermediate in the production of polyesters. It also used in essence, perfumes and in pharmaceuticals. The diacrylate is hydrophobic and used for flexible dental materials, radiation curable coatings, and for lenses.

Solubility
Soluble in alcohol. Insoluble in water, petroleum ether.

Notes
Store in a cool, dry, well-ventilated area. Incompatible with oxidizing agent.

Chemical Identifiers

CAS 112-47-0
Molecular Formula C10H22O2
Molecular Weight (g/mol) 174.284
MDL Number MFCD00004749
InChI Key FOTKYAAJKYLFFN-UHFFFAOYSA-N
Synonym 1,10-decanediol, decamethylene glycol, decamethylenediol, 1,10-dihydroxydecane, 1,10-decamethylenediol, 1,10-decamethylene diol, 1,6-bis 2-hydroxyethyl hexane, alpha,omega-decanediol, unii-5i577udk52, 1,10-ddecanediol
PubChem CID 37153
IUPAC Name decane-1,10-diol
SMILES C(CCCCCO)CCCCO

Specifications

Melting Point 71°C to 74°C
Density 1.08
Boiling Point 297°C
Flash Point 152°C (306°F)
Quantity 100 g
Beilstein 1698975
Merck Index 14,2849
Solubility Information Soluble in alcohol. Insoluble in water,petroleum ether.
Formula Weight 174.28
Percent Purity 97%
Chemical Name or Material 1,10-Decanediol
Show More Show Less
EINECSNumber 203-975-2
RTECSNumber HD8433713
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only