missing translation for 'onlineSavingsMsg'
Learn More

1,1'-Thiocarbonyldiimidazole 95.0+%, TCI America™

Catalog No. c137625g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
C137625G 25 g
C13765G 5 g
2 options

Catalog No. C137625G

Supplier: TCI America C137625G

Only null left

Chemical Identifiers

CAS 6160-65-2
Molecular Formula C7H6N4S
Molecular Weight (g/mol) 178.21
MDL Number MFCD00005289
InChI Key RAFNCPHFRHZCPS-UHFFFAOYSA-N
Synonym 1,1'-thiocarbonyldiimidazole, thiocarbonyldiimidazole, n,n'-thiocarbonyldiimidazole, di 1h-imidazol-1-yl methanethione, 1,1-thiocarbonyldiimidazole, di imidazol-1-yl methanethione, n,n'-thiocarbonyldiimidazole tcdi, 1h-imidazole, 1,1'-carbonothioylbis, n,n-thiocarbonyldiimidazole, 1,1'-carbonothioylbis-1h-imidazole
PubChem CID 80264
IUPAC Name 1-(1H-imidazole-1-carbothioyl)-1H-imidazole
SMILES S=C(N1C=CN=C1)N1C=CN=C1

Specifications

Melting Point 102°C
Quantity 25 g
Formula Weight 178.21
Percent Purity ≥95.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,1′-Thiocarbonyldiimidazole
TSCA No
Recommended Storage Refrigerator