CAS RN 6160-65-2
IUPAC Name:
1-(1H-imidazole-1-carbothioyl)-1H-imidazole
Synonyms:
thiocarbonyldiimidazole
Molecular Weight (g/mol):
178.21
Molecular Formula:
C7H6N4S
InChi Key:
RAFNCPHFRHZCPS-UHFFFAOYSA-N
SMILES:
S=C(N1C=CN=C1)N1C=CN=C1
1
–
1
of
1
results
1
1,1'-Thiocarbonyldiimidazole 95.0+%, TCI America™
CAS: 6160-65-2 Molecular Formula: C7H6N4S Molecular Weight (g/mol): 178.21 MDL Number: MFCD00005289 InChI Key: RAFNCPHFRHZCPS-UHFFFAOYSA-N Synonym: 1,1'-thiocarbonyldiimidazole,thiocarbonyldiimidazole,n,n'-thiocarbonyldiimidazole,di 1h-imidazol-1-yl methanethione,1,1-thiocarbonyldiimidazole,di imidazol-1-yl methanethione,n,n'-thiocarbonyldiimidazole tcdi,1h-imidazole, 1,1'-carbonothioylbis,n,n-thiocarbonyldiimidazole,1,1'-carbonothioylbis-1h-imidazole PubChem CID: 80264 IUPAC Name: 1-(1H-imidazole-1-carbothioyl)-1H-imidazole SMILES: S=C(N1C=CN=C1)N1C=CN=C1