missing translation for 'onlineSavingsMsg'
Learn More

1,1'-Thiocarbonyldiimidazole, 90%, Tech.

Catalog No. p-3737500
Change view
Click to view available options
Quantity:
10 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC168480100 10 g Glass bottle
AC168480250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC168480100 Supplier Thermo Scientific Chemicals Supplier No. 168480100
Only null left

CAS: 6160-65-2 | C7H6N4S | 178.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6160-65-2
Molecular Formula C7H6N4S
Molecular Weight (g/mol) 178.21
MDL Number MFCD00005289
InChI Key RAFNCPHFRHZCPS-UHFFFAOYSA-N
Synonym 1,1'-thiocarbonyldiimidazole, thiocarbonyldiimidazole, n,n'-thiocarbonyldiimidazole, di 1h-imidazol-1-yl methanethione, 1,1-thiocarbonyldiimidazole, di imidazol-1-yl methanethione, n,n'-thiocarbonyldiimidazole tcdi, 1h-imidazole, 1,1'-carbonothioylbis, n,n-thiocarbonyldiimidazole, 1,1'-carbonothioylbis-1h-imidazole
PubChem CID 80264
IUPAC Name di(imidazol-1-yl)methanethione
SMILES S=C(N1C=CN=C1)N1C=CN=C1

Specifications

CAS Min % 88.0
CAS Max % 100.0
Melting Point 96.0°C to 103.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 90%
Packaging Glass bottle
Quantity 10 g
Fieser 01,1151; 02,411; 05,661; 06,583; 13,318
Solubility Information Solubility in water: decomposes.
Formula Weight 178.21
Percent Purity 90%
Grade Technical
Physical Form Powder
Chemical Name or Material 1, 1'-Thiocarbonyldiimidazole
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 228-183-4

RUO – Research Use Only