missing translation for 'onlineSavingsMsg'
Learn More

1,1'-Thiocarbonyldi-2(1H)-pyridone 98.0+%, TCI America™

Catalog No. p-7137824
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T36441G 1 g
T36445G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T36441G Supplier TCI America Supplier No. T36441G
Only null left

Chemical Identifiers

CAS 102368-13-8
Molecular Formula C11H8N2O2S
Molecular Weight (g/mol) 232.26
MDL Number MFCD00075238
InChI Key KXMMNJQMGILZDB-UHFFFAOYSA-N
PubChem CID 853162
IUPAC Name 1-(2-oxo-1,2-dihydropyridine-1-carbothioyl)-1,2-dihydropyridin-2-one
SMILES O=C1C=CC=CN1C(=S)N1C=CC=CC1=O

Specifications

Melting Point 158°C
Quantity 1 g
Synonym 1,1'-thiocarbonyldi-2 1h-pyridone, 1,1'-thiocarbonylbis pyridin-2 1h-one, 2 1h-pyridinone, 1,1'-carbonothioylbis, 2 1h-pyridinone,1,1'-carbonothioylbis, 1-2-oxopyridine-1-carbothioyl pyridin-2-one, acmc-20a80d, 1,1'-thiocarbonyldi-2-pyridone, 1,1-thiocarbonyldi-2 1h-pyridone, 1,1'-thiocarbonyldi-2 2h-pyridone, 1,'1-thiocarbonyldi-2 1h-pyridone
Formula Weight 232.26
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 1,1′-Thiocarbonyldi-2(1H)-pyridone