accessibility menu, dialog, popup

UserName

ONSNO

CAS RN 102368-13-8

IUPAC Name: 1-(2-oxo-1,2-dihydropyridine-1-carbothioyl)-1,2-dihydropyridin-2-one
Synonyms: 1-(2-oxopyridine-1-carbothioyl)pyridin-2-one
Molecular Weight (g/mol): 232.26
Molecular Formula: C11H8N2O2S
InChi Key: KXMMNJQMGILZDB-UHFFFAOYSA-N
SMILES: O=C1C=CC=CN1C(=S)N1C=CC=CC1=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Grade 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)