missing translation for 'onlineSavingsMsg'
Learn More

1,1'-Thiocarbonyldi-2(1H)-pyridone, 95%

Catalog No. AAH3008703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3008703 1 g
AAH3008706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3008703 Supplier Thermo Scientific Chemicals Supplier No. H3008703
Only null left

CAS: 102368-13-8 | C11H8N2O2S | 232.26 g/mol

1,1'-Thiocarbonyldi-2(1H)-pyridone is used in the preparation of thio-analogs of thioureas, sulforaphane and 2-furan-2-yl-3-hydroxy-6-isothiocyanato-chromen-4-one. It is involved in the preparation of norbiotinamine and its reactive derivatives such as isothiocyanates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1′-Thiocarbonyldi-2(1H)-pyridone is used in the preparation of thio-analogs of thioureas, sulforaphane and 2-furan-2-yl-3-hydroxy-6-isothiocyanato-chromen-4-one. It is involved in the preparation of norbiotinamine and its reactive derivatives such as isothiocyanates.

Notes
Incompatible with strong oxidizing agents. Store in a cool place.

Chemical Identifiers

CAS 102368-13-8
Molecular Formula C11H8N2O2S
Molecular Weight (g/mol) 232.26
MDL Number MFCD00075238
InChI Key KXMMNJQMGILZDB-UHFFFAOYSA-N
PubChem CID 853162
IUPAC Name 1-(2-oxopyridine-1-carbothioyl)pyridin-2-one
SMILES O=C1C=CC=CN1C(=S)N1C=CC=CC1=O

Specifications

Melting Point 163°C to 166°C
Quantity 1 g
Synonym 1,1'-thiocarbonyldi-2 1h-pyridone, 1,1'-thiocarbonylbis pyridin-2 1h-one, 2 1h-pyridinone, 1,1'-carbonothioylbis, 2 1h-pyridinone,1,1'-carbonothioylbis, 1-2-oxopyridine-1-carbothioyl pyridin-2-one, acmc-20a80d, 1,1'-thiocarbonyldi-2-pyridone, 1,1-thiocarbonyldi-2 1h-pyridone, 1,1'-thiocarbonyldi-2 2h-pyridone, 1,'1-thiocarbonyldi-2 1h-pyridone
Formula Weight 232.26
Percent Purity 95%
Chemical Name or Material 1,1'-Thiocarbonyldi-2(1H)-pyridone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only