missing translation for 'onlineSavingsMsg'
Learn More

1,1'-Thiobis(2-naphthol) 98.0+%, TCI America™

Catalog No. T100425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T100425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T100425G Supplier TCI America Supplier No. T100425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 17096-15-0
Molecular Formula C20H14O2S
Molecular Weight (g/mol) 318.39
MDL Number MFCD00046469
InChI Key HGYMQZVPTMKXGI-UHFFFAOYSA-N
Synonym Bis(2-hydroxy-1-naphthyl) Sulfide, 1,1′C-Thiodi(2-naphthol)
PubChem CID 86951
IUPAC Name 1-(2-hydroxynaphthalen-1-yl)sulfanylnaphthalen-2-ol
SMILES C1=CC=C2C(=C1)C=CC(=C2SC3=C(C=CC4=CC=CC=C43)O)O

Specifications

Melting Point 216°C
Quantity 25 g
Formula Weight 318.39
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,1′-Thiobis(2-naphthol)