missing translation for 'onlineSavingsMsg'
Learn More

1-(1-Naphthyl)-2-(trimethylsilyl)acetylene, 97%

Catalog No. p-7052813
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5595303 1 g
AAH5595306 5 g
AAH5595314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH5595303 Supplier Thermo Scientific Chemicals Supplier No. H5595303
Only null left

CAS: 104784-51-2 | C15H16Si | 224.378 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 104784-51-2
Molecular Formula C15H16Si
Molecular Weight (g/mol) 224.378
MDL Number MFCD04039886
InChI Key WATBCTJRCLNXNF-UHFFFAOYSA-N
Synonym 1-1-naphthyl-2-trimethylsilyl acetylene, trimethyl 2-naphthalen-1-ylethynyl silane, trimethyl 2-naphthalen-1-yl ethynyl silane, 1-naphthylethynyl trimethylsilane, 1-trimethylsilylethynyl naphthalene, 1-trimethylsilyl ethynyl naphthalene, trimethyl naphthalen-1-ylethynyl silane, trimethyl naphthalen-1-yl ethynyl silane, trimethylnaphthalen-1-ylethynylsilane
PubChem CID 4438232
IUPAC Name trimethyl(2-naphthalen-1-ylethynyl)silane
SMILES C[Si](C)(C)C#CC1=CC=CC2=CC=CC=C21

Specifications

Melting Point 41°C to 43°C
Density 0.968 g/mL
Boiling Point 292°C to 293°C
Flash Point >110°C (230°F)
Refractive Index 1.595
Quantity 1 g
Formula Weight 224.38
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-(1-Naphthyl)-2-(trimethylsilyl)acetylene
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only