accessibility menu, dialog, popup

UserName

CAS RN 104784-51-2

IUPAC Name: trimethyl[2-(naphthalen-1-yl)ethynyl]silane
Synonyms: trimethyl[2-(naphthalen-1-yl)ethynyl]silane
Molecular Weight (g/mol): 224.38
Molecular Formula: C15H16Si
InChi Key: WATBCTJRCLNXNF-UHFFFAOYSA-N
SMILES: C[Si](C)(C)C#CC1=C2C=CC=CC2=CC=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)