missing translation for 'onlineSavingsMsg'
Learn More

1-(1-Methyl-4-piperidinyl)piperazine, 98%

Catalog No. AAH2762406
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2762406 5 g
AAH2762403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2762406 Supplier Thermo Scientific Chemicals Supplier No. H2762406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 23995-88-2 | C10H21N3 | 183.299 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 23995-88-2
Molecular Formula C10H21N3
Molecular Weight (g/mol) 183.299
MDL Number MFCD01075186
InChI Key OHUMKYGINIODOY-UHFFFAOYSA-N
Synonym 1-1-methyl-4-piperidinyl piperazine, 1-1-methylpiperidin-4-yl piperazine, 1-n-methylpiperidin-4-yl piperazine, 1-1-methyl-piperidin-4-yl-piperazine, piperazine, 1-1-methyl-4-piperidinyl, 1-1-methyl-4-piperidyl piperazine, 1-1-methyl-4-piperidino piperazine, zlchem 509, acmc-1cq7x, 1-methyl-4-piperidyl piperazine
PubChem CID 566324
IUPAC Name 1-(1-methylpiperidin-4-yl)piperazine
SMILES CN1CCC(CC1)N2CCNCC2

Specifications

Melting Point 29°C to 32°C
Refractive Index 1.508
Quantity 5 g
UN Number UN3259
Sensitivity Air Sensitive
Formula Weight 183.3
Percent Purity 98%
Chemical Name or Material 1-(1-Methyl-4-piperidinyl)piperazine
Hazard Category H302+H312+H332-H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only