CAS RN 23995-88-2
IUPAC Name:
1-(1-methylpiperidin-4-yl)piperazine
Synonyms:
1-(1-methylpiperidin-4-yl)piperazine
Molecular Weight (g/mol):
183.3
Molecular Formula:
C10H21N3
InChi Key:
OHUMKYGINIODOY-UHFFFAOYSA-N
SMILES:
CN1CCC(CC1)N1CCNCC1
1
–
1
of
1
results
1
1-(1-Methyl-4-piperidinyl)piperazine, 98%
CAS: 23995-88-2 Molecular Formula: C10H21N3 Molecular Weight (g/mol): 183.299 MDL Number: MFCD01075186 InChI Key: OHUMKYGINIODOY-UHFFFAOYSA-N Synonym: 1-1-methyl-4-piperidinyl piperazine,1-1-methylpiperidin-4-yl piperazine,1-n-methylpiperidin-4-yl piperazine,1-1-methyl-piperidin-4-yl-piperazine,piperazine, 1-1-methyl-4-piperidinyl,1-1-methyl-4-piperidyl piperazine,1-1-methyl-4-piperidino piperazine,zlchem 509,acmc-1cq7x,1-methyl-4-piperidyl piperazine PubChem CID: 566324 IUPAC Name: 1-(1-methylpiperidin-4-yl)piperazine SMILES: CN1CCC(CC1)N2CCNCC2