missing translation for 'onlineSavingsMsg'
Learn More

1,1-Diphenylpropane, 98+%

Catalog No. AAL0927406
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0927406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0927406 Supplier Thermo Scientific Chemicals Supplier No. L0927406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1530-03-6 | C15H16 | 196.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1530-03-6
Molecular Formula C15H16
Molecular Weight (g/mol) 196.29
MDL Number MFCD00041670
InChI Key BUZMJVBOGDBMGI-UHFFFAOYSA-N
Synonym 1,1-diphenylpropane, diphenylpropane, benzene, 1,1'-propylidenebis, propane, diphenyl, 1,1-diphenylpropyl, phenylpropyl benzene, 3,3-diphenylpropane, acmc-1cujs, 1-phenylpropyl benzene #
PubChem CID 73726
IUPAC Name 1-phenylpropylbenzene
SMILES CCC(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Density 0.995
Refractive Index 1.564
Quantity 5 g
Beilstein 2044542
Formula Weight 196.29
Percent Purity 99%
Chemical Name or Material 1,1-Diphenylpropane
EINECSNumber 216-222-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only