missing translation for 'onlineSavingsMsg'
Learn More

1,1-Diphenylhydrazine hydrochloride, 98%

Catalog No. AAA1128906
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1128906 5 g
AAA1128914 25 g
AAA1128922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1128906 Supplier Thermo Scientific Chemicals Supplier No. A1128906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 530-47-2 | C12H13ClN2 | 220.7 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 530-47-2
Molecular Formula C12H13ClN2
Molecular Weight (g/mol) 220.7
MDL Number MFCD00012950
InChI Key MIVUDWFNUOXEJM-UHFFFAOYSA-N
Synonym 1,1-diphenylhydrazine hydrochloride, n,n-diphenylhydrazine hydrochloride, unii-o8100cbz4n, diphenyl hydrazine hydrochloride, hydrazine, 1,1-diphenyl-, monohydrochloride, n,n-diphenylhydrazinium 1+ chloride, 1,1-diphenylhydrazine hcl, 1,1-diphenylhydrazine monohydrochloride, aminodiphenylamine, chloride, hydrazine, 1,1-diphenyl-, hydrochloride 1:1
PubChem CID 10738
IUPAC Name 1,1-diphenylhydrazine;hydrochloride
SMILES C1=CC=C(C=C1)N(C2=CC=CC=C2)N.Cl

Specifications

Melting Point ∼163°C (decomposition)
Odor Odorless
Quantity 5 g
Beilstein 3569340
Merck Index 14,3325
Formula Weight 220.7
Percent Purity 98%
Chemical Name or Material 1,1-Diphenylhydrazine hydrochloride
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 208-481-0
RTECSNumber MV3520000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only