missing translation for 'onlineSavingsMsg'
Learn More

1,1-Diphenylethylene (stabilized with HQ) 98.0+%, TCI America™

Catalog No. p-7126484
Change view
Click to view available options
Quantity:
5 g
25 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D08865G 5 g
D088625G 25 g
D0886250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D08865G Supplier TCI America Supplier No. D08865G
Only null left

Chemical Identifiers

CAS 530-48-3
Molecular Formula C14H12
Molecular Weight (g/mol) 180.25
MDL Number MFCD00008583
InChI Key ZMYIIHDQURVDRB-UHFFFAOYSA-N
Synonym 1,1-diphenylethylene, ethene-1,1-diyldibenzene, 1,1-diphenylethene, benzene, 1,1'-ethenylidenebis, 1-phenylethenyl benzene, unii-bx0l5b6lll, as-diphenylethylene, .alpha.-phenylstyrene, 1-phenylvinyl benzene, bx0l5b6lll
PubChem CID 10740
IUPAC Name 1-phenylethenylbenzene
SMILES C=C(C1=CC=CC=C1)C2=CC=CC=C2

Specifications

Color Yellow
Boiling Point 277°C
Quantity 5 g
Formula Weight 180.25
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,1-Diphenylethylene (stabilized with HQ)
TSCA Yes
Recommended Storage Refrigerator