missing translation for 'onlineSavingsMsg'
Learn More

1,1-Diphenylethylene, 98%, Thermo Scientific™

$105.35 - $809.96

Chemical Identifiers

CAS 530-48-3
Molecular Formula C14H12
Molecular Weight (g/mol) 180.25
InChI Key ZMYIIHDQURVDRB-UHFFFAOYSA-N
Synonym 1,1-diphenylethylene, ethene-1,1-diyldibenzene, 1,1-diphenylethene, benzene, 1,1'-ethenylidenebis, 1-phenylethenyl benzene, unii-bx0l5b6lll, as-diphenylethylene, .alpha.-phenylstyrene, 1-phenylvinyl benzene, bx0l5b6lll
PubChem CID 10740
IUPAC Name 1-phenylethenylbenzene
SMILES C=C(C1=CC=CC=C1)C2=CC=CC=C2
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC117260100
SDS View Documents
Thermo Scientific Chemicals
117260100
10g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC117260250
SDS View Documents
Thermo Scientific Chemicals
117260250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC117261000
SDS View Documents
Thermo Scientific Chemicals
117261000
100g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

530-48-3
180.25
1,1-diphenylethylene, ethene-1,1-diyldibenzene, 1,1-diphenylethene, benzene, 1,1'-ethenylidenebis, 1-phenylethenyl benzene, unii-bx0l5b6lll, as-diphenylethylene, .alpha.-phenylstyrene, 1-phenylvinyl benzene, bx0l5b6lll
1-phenylethenylbenzene
C14H12
ZMYIIHDQURVDRB-UHFFFAOYSA-N
10740
C=C(C1=CC=CC=C1)C2=CC=CC=C2

Specifications

530-48-3
100.0
270.0°C to 271.0°C
Authentic
Glass bottle
1.6065 to 1.6085
10g
1.02
1,1-diphenylethylene, ethene-1,1-diyldibenzene, 1,1-diphenylethene, benzene, 1,1'-ethenylidenebis, 1-phenylethenyl benzene, unii-bx0l5b6lll, as-diphenylethylene, .alpha.-phenylstyrene, 1-phenylvinyl benzene, bx0l5b6lll
C=C(C1=CC=CC=C1)C2=CC=CC=C2
180.25
180.25
1, 1-Diphenylethylene
97.5
1.0200g/mL
>112°C
97.5% min. (GC)
C14H12
(C6H5)2C=CH2
05,639
15,3356
ZMYIIHDQURVDRB-UHFFFAOYSA-N
1-phenylethenylbenzene
10740
98%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 208-482-6

SDS
Documents

Documents

RUO – Research Use Only