missing translation for 'onlineSavingsMsg'
Learn More

1,1-Diphenylethylene, 98%

Catalog No. aaa1443430
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
250 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1443430 250 g
AAA1443409 10 g
AAA1443414 25 g
AAA1443422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA1443430 Supplier Thermo Scientific Chemicals Supplier No. A1443430
Only null left

1,1-Diphenylethylene, 98%, C14H12, CAS Number-530-48-3, unii-bx0l5b6lll, 1,1-diphenylethene, 1,1-diphenylethylene, 1-phenylethenyl benzene, 1-phenylvinyl benzene, ethene-1,1-diyldibenzene, bx0l5b6lll, .alpha.-phenylstyrene, benzene, 1,1'-ethenylidenebis, as-diphenylethylene, 250g, 1099062 | CAS: 530-48-3 | C14H12 | 180.25 g/mol

1,1-Diphenylethylene is used in the preparation of 2-chloro-1,1-diphenyl-ethene by reacting with benzeneseleninyl chloride and aluminum(III) chloride as reagents. It acts as an intermediate in organic synthesis as well as in pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1-Diphenylethylene is used in the preparation of 2-chloro-1,1-diphenyl-ethene by reacting with benzeneseleninyl chloride and aluminum(III) chloride as reagents. It acts as an intermediate in organic synthesis as well as in pharmaceuticals.

Solubility
Miscible with methanol, chloroform and ether. Insoluble in water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents

Chemical Identifiers

CAS 530-48-3
Molecular Formula C14H12
Molecular Weight (g/mol) 180.25
MDL Number MFCD00008583
InChI Key ZMYIIHDQURVDRB-UHFFFAOYSA-N
Synonym 1,1-diphenylethylene, ethene-1,1-diyldibenzene, 1,1-diphenylethene, benzene, 1,1'-ethenylidenebis, 1-phenylethenyl benzene, unii-bx0l5b6lll, as-diphenylethylene, .alpha.-phenylstyrene, 1-phenylvinyl benzene, bx0l5b6lll
PubChem CID 10740
IUPAC Name 1-phenylethenylbenzene
SMILES C=C(C1=CC=CC=C1)C2=CC=CC=C2

Specifications

Melting Point 5°C to 7°C
Density 1.023
Boiling Point 270°C to 271°C
Flash Point 136°C (276°F)
Refractive Index 1.6065
Quantity 250 g
Beilstein 1099062
Merck Index 14,3323
Solubility Information Miscible with methanol,chloroform and ether. Insoluble in water.
Formula Weight 180.25
Percent Purity 98%
Chemical Name or Material 1,1-Diphenylethylene
Show More Show Less
Hazard Category H317
Precautionary Statement P261-P272-P280-P302+P352-P333+P313-P363-P501c
EINECSNumber 208-482-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only