missing translation for 'onlineSavingsMsg'
Learn More

1,1-Diphenyl-2-propyn-1-ol 97.0+%, TCI America™

Catalog No. p-7127334
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
D24955G 5 g
D249525G 25 g
2 options

Catalog No. D24955G

Supplier: TCI America D24955G

Only null left

Chemical Identifiers

CAS 3923-52-2
Molecular Formula C15H12O
Molecular Weight (g/mol) 208.26
MDL Number MFCD00041570
InChI Key SMCLTAARQYTXLW-UHFFFAOYSA-N
Synonym 1,1-diphenyl-2-propyn-1-ol, diphenylethinylcarbinol, ethynyldiphenylmethanol, 1,1-diphenylpropargyl alcohol, diphenylethynylcarbinol, 3,3-diphenyl-3-hydroxy-1-propyne, benzenemethanol, .alpha.-ethynyl-.alpha.-phenyl, 2-propyn-1-ol, 1,1-diphenyl, benzenemethanol, alpha-ethynyl-alpha-phenyl, benzenemethanol, a-ethynyl-a-phenyl
PubChem CID 92976
IUPAC Name 1,1-diphenylprop-2-yn-1-ol
SMILES OC(C#C)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 48°C
Color White-Yellow
Boiling Point 200°C
Quantity 5 g
Formula Weight 208.26
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,1-Diphenyl-2-propyn-1-ol
RTECSNumber DA4796050
TSCA No