missing translation for 'onlineSavingsMsg'
Learn More

1,1-Diphenyl-2-propyn-1-ol, 99%

Catalog No. p-7025766
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2142806 5 g
AAB2142814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2142806 Supplier Thermo Scientific Chemicals Supplier No. B2142806
Only null left

CAS: 3923-52-2 | C15H12O | 208.26 g/mol

1,1-Diphenyl-2-propyn-1-ol is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is used in medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1-Diphenyl-2-propyn-1-ol is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is used in medicine.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 3923-52-2
Molecular Formula C15H12O
Molecular Weight (g/mol) 208.26
MDL Number MFCD00041570
InChI Key SMCLTAARQYTXLW-UHFFFAOYSA-N
Synonym 1,1-diphenyl-2-propyn-1-ol, diphenylethinylcarbinol, ethynyldiphenylmethanol, 1,1-diphenylpropargyl alcohol, diphenylethynylcarbinol, 3,3-diphenyl-3-hydroxy-1-propyne, benzenemethanol, .alpha.-ethynyl-.alpha.-phenyl, 2-propyn-1-ol, 1,1-diphenyl, benzenemethanol, alpha-ethynyl-alpha-phenyl, benzenemethanol, a-ethynyl-a-phenyl
PubChem CID 92976
IUPAC Name 1,1-diphenylprop-2-yn-1-ol
SMILES OC(C#C)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 44°C to 48°C
Boiling Point 179°C to 180°C (18 mmHg)
Flash Point 113°C (235°F)
Quantity 5 g
Beilstein 514549
Solubility Information Insoluble in water.
Formula Weight 208.26
Percent Purity 99%
Chemical Name or Material 1,1-Diphenyl-2-propyn-1-ol
Hazard Category H315-H320-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
RTECSNumber DA4796050
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only