missing translation for 'onlineSavingsMsg'
Learn More

1,1-Dimethylpiperidinium Chloride 98.0+%, TCI America™

Catalog No. D40165G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D40165G 5 g
D40161G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D40165G Supplier TCI America Supplier No. D40165G

Chemical Identifiers

CAS 24307-26-4
Molecular Formula C7H16ClN
Molecular Weight (g/mol) 149.662
MDL Number MFCD00055543
InChI Key VHOVSQVSAAQANU-UHFFFAOYSA-M
Synonym Mepiquat Chloride
PubChem CID 62781
ChEBI CHEBI:81772
IUPAC Name 1,1-dimethylpiperidin-1-ium;chloride
SMILES C[N+]1(CCCCC1)C.[Cl-]

Specifications

Color White-Yellow
pH 6.74
Quantity 5 g
Formula Weight 149.66
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,1-Dimethylpiperidinium Chloride
RTECSNumber TN4939200
TSCA Yes