accessibility menu, dialog, popup

UserName

ClCH3NH3C

CAS RN 24307-26-4

IUPAC Name: 1,1-dimethylpiperidin-1-ium chloride
Synonyms: mepiquat chloride
Molecular Weight (g/mol): 149.66
Molecular Formula: C7H16ClN
InChi Key: VHOVSQVSAAQANU-UHFFFAOYSA-M
SMILES: [Cl-].C[N+]1(C)CCCCC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results