missing translation for 'onlineSavingsMsg'
Learn More

1,1-Dimethoxycyclopentane 96.0+%, TCI America™

Catalog No. p-7126991
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D18861ML 1 mL
D18865ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D18861ML Supplier TCI America Supplier No. D18861ML

Chemical Identifiers

CAS 931-94-2
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.187
MDL Number MFCD00060058
InChI Key AGWFDZMDKNQQHG-UHFFFAOYSA-N
Synonym Cyclopentanone Dimethylketal
PubChem CID 523052
IUPAC Name 1,1-dimethoxycyclopentane
SMILES COC1(CCCC1)OC

Specifications

Color Colorless
Boiling Point 140°C
Quantity 1 mL
UN Number 3271
Formula Weight 130.19
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,1-Dimethoxycyclopentane