missing translation for 'onlineSavingsMsg'
Learn More

1,1'-Difluoro-2,2'-bipyridinium Bis(tetrafluoroborate) 95.0+%, TCI America™

Catalog No. D38121G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D38121G 1 g
D38125G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D38121G Supplier TCI America Supplier No. D38121G

Chemical Identifiers

CAS 178439-26-4
Molecular Formula C10H8B2F10N2
Molecular Weight (g/mol) 367.792
MDL Number MFCD01073546
InChI Key IDFABAACDRPWQX-UHFFFAOYSA-N
Synonym 1,1'-difluoro-2,2'-bipyridinium bis tetrafluoroborate, 1,1'-difluoro-2,2'-bipyridine-1,1'-diium tetrafluoroborate, n,n'-difluoro-2,2'-bipyridinium bis tetrafluoroborate, 1-fluoro-2-1-fluoropyridin-1-ium-2-yl pyridin-1-ium ditetrafluoroborate, mec-31, synfluor reagent, 1,1'-difluoro-2,2'-bipyridine-1,1'-diium ditetrafluoroborate, daikin mec-31, acmc-209eei, n,n'-difluoro-2,2'-bipyridinium
PubChem CID 2733931
IUPAC Name 1-fluoro-2-(1-fluoropyridin-1-ium-2-yl)pyridin-1-ium;ditetrafluoroborate
SMILES [B-](F)(F)(F)F.[B-](F)(F)(F)F.C1=CC=[N+](C(=C1)C2=CC=CC=[N+]2F)F

Specifications

Color White-Yellow
Quantity 1 g
UN Number 1759
Formula Weight 367.79
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,1′-Difluoro-2,2′-bipyridinium Bis(tetrafluoroborate)
TSCA No
Recommended Storage Refrigerator