missing translation for 'onlineSavingsMsg'
Learn More

1,1'-Diethyl-4,4'-carbocyanine iodide, 96%

Catalog No. p-7052712
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH55456MD 250 mg
AAH5545603 1 g
AAH5545606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH55456MD Supplier Thermo Scientific Chemicals Supplier No. H55456MD
Only null left

CAS: 4727-50-8 | C25H25IN2 | 480.393 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4727-50-8
Molecular Formula C25H25IN2
Molecular Weight (g/mol) 480.393
MDL Number MFCD00011970
InChI Key CEJANLKHJMMNQB-UHFFFAOYSA-M
Synonym cryptocyanin, cryptocyanine, kryptocyanin, kryptocyanine, cryptocyanine iodide, kryptocyanine iodide, 1,1'-diethylquino-4-carbocyanine iodide, cryptocyanine o.a. 2, 1,4'-carbocyanine iodide, 1,4'-quinocarbocyanine iodide
PubChem CID 16219292
ChEBI CHEBI:51502
IUPAC Name (4E)-1-ethyl-4-[(Z)-3-(1-ethylquinolin-1-ium-4-yl)prop-2-enylidene]quinoline;iodide
SMILES CCN1C=CC(=CC=CC2=CC=[N+](C3=CC=CC=C23)CC)C4=CC=CC=C41.[I-]

Specifications

Melting Point 250°C to 253°C
Color Green
Quantity 250 mg
UN Number UN2811
Sensitivity Light sensitive
Formula Weight 480.38
Percent Purity 96%
Physical Form Powder
Chemical Name or Material 1,1'-Diethyl-4,4'-carbocyanine iodide
Hazard Category H300-H311+H331-H315-H319
Hazard Statement GHS H Statement
H300-H315-H319-H335
Fatal if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 225-224-8
RTECSNumber VC3676880
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only