missing translation for 'onlineSavingsMsg'
Learn More

1,1-Dichlorodimethyl ether, 97%

Catalog No. p-3757597
Change view
Click to view available options
Quantity:
10 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC113470100 10 g Glass bottle
AC113471000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC113470100 Supplier Thermo Scientific Chemicals Supplier No. 113470100
Only null left

CAS: 4885-02-3 | C2H4Cl2O | 114.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4885-02-3
Molecular Formula C2H4Cl2O
Molecular Weight (g/mol) 114.96
MDL Number MFCD00000828
InChI Key GRTGGSXWHGKRSB-UHFFFAOYSA-N
SMILES COC(Cl)Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.2700g/mL
Boiling Point 85.0°C
Flash Point 22°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula Cl2CHOCH3
Refractive Index 1.4295 to 1.4325
Quantity 10 g
Fieser 01,220; 02,120; 05,200; 06,171; 07,98; 08,154
Specific Gravity 1.27
Synonym dichloromethyl methyl ether, 1,1-dichlorodimethyl ether, dichloro methoxy methane, methane, dichloromethoxy, ether, dichloromethyl methyl, alpha,alpha-dichloromethyl methyl ether, methoxydichloromethane, 1,1-dichloromethyl methyl ether, alpha,alpha-dichloromethyl ether, dichloromethylmethylether
IUPAC Name dichloro(methoxy)methane
PubChem CID 21004
Formula Weight 114.96
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1, 1-Dichlorodimethyl ether
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Highly flammable liquid and vapor.
Harmful if inhaled.
In use,may form flammable/explosive vapor-air mixture.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Ground/Bond container and receiving equipment.
IF IN EYES: Rinse cautiously with water
EINECSNumber 225-498-9

RUO – Research Use Only