missing translation for 'onlineSavingsMsg'
Learn More

1,1'-Dibutyl-3,3,3',3'-tetramethylindotricarbocyanine Hexafluorophosphate 98.0+%, TCI America™

Catalog No. p-7128608
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D50131G 1 g
D50135G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D50131G Supplier TCI America Supplier No. D50131G
Only null left

Chemical Identifiers

CAS 134339-08-5
Molecular Formula C35H45F6N2P
Molecular Weight (g/mol) 638.72
MDL Number MFCD27966053
InChI Key GUFQKTNQEKEMJE-UHFFFAOYSA-N
Synonym 1-Butyl-2-[7-(1-butyl-1,3-dihydro-3,3-dimethyl-2H-indol-2-ylidene)-1,3,5-heptatrien-1-yl]-3,3-dimethyl-3H-indolium Hexafluorophosphate
PubChem CID 133556290
IUPAC Name 1-butyl-2-[7-(1-butyl-3,3-dimethyl-2,3-dihydro-1H-indol-2-ylidene)hepta-1,3,5-trien-1-yl]-3,3-dimethyl-3H-indol-1-ium; hexafluoro-λ⁵-phosphanuide
SMILES F[P-](F)(F)(F)(F)F.CCCCN1C2=CC=CC=C2C(C)(C)C1=CC=CC=CC=CC1=[N+](CCCC)C2=CC=CC=C2C1(C)C

Specifications

Quantity 1 g
Formula Weight 638.72
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1,1′-Dibutyl-3,3,3′,3′-tetramethylindotricarbocyanine Hexafluorophosphate