accessibility menu, dialog, popup

UserName

CH3NNCH3H3CH3CH3CH3CFPFFFFF(E/Z)(E/Z)(E/Z)(E/Z)

CAS RN 134339-08-5

IUPAC Name: 1-butyl-2-[7-(1-butyl-3,3-dimethyl-2,3-dihydro-1H-indol-2-ylidene)hepta-1,3,5-trien-1-yl]-3,3-dimethyl-3H-indol-1-ium; hexafluoro-λ⁵-phosphanuide
Synonyms: 1-butyl-2-[7-(1-butyl-3,3-dimethylindol-2-ylidene)hepta-1,3,5-trien-1-yl]-3,3-dimethylindol-1-ium hexafluorophosphate
Molecular Weight (g/mol): 638.72
Molecular Formula: C35H45F6N2P
InChi Key: GUFQKTNQEKEMJE-UHFFFAOYSA-N
SMILES: F[P-](F)(F)(F)(F)F.CCCCN1C2=CC=CC=C2C(C)(C)C1=CC=CC=CC=CC1=[N+](CCCC)C2=CC=CC=C2C1(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results