missing translation for 'onlineSavingsMsg'
Learn More

1,1-Dibromo-2,2-bis(chloromethyl)cyclopropane 96.0+%, TCI America™

Catalog No. D52901G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D52901G 1 g
D52905G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D52901G Supplier TCI America Supplier No. D52901G
Only null left

Chemical Identifiers

CAS 98577-44-7
Molecular Formula C5H6Br2Cl2
Molecular Weight (g/mol) 296.81
MDL Number MFCD00101445
InChI Key RCRVZCIUKQNOIS-UHFFFAOYSA-N
PubChem CID 534192
IUPAC Name 1,1-dibromo-2,2-bis(chloromethyl)cyclopropane
SMILES ClCC1(CCl)CC1(Br)Br

Specifications

Melting Point 50°C
Color White-Yellow
Quantity 1 g
Formula Weight 296.81
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,1-Dibromo-2,2-bis(chloromethyl)cyclopropane