CAS RN 98577-44-7
CAS RN 98577-44-7
IUPAC Name:
1,1-dibromo-2,2-bis(chloromethyl)cyclopropane
Synonyms:
1,1-dibromo-2,2-bis(chloromethyl)cyclopropane
Molecular Weight (g/mol):
296.81
Molecular Formula:
C5H6Br2Cl2
InChi Key:
RCRVZCIUKQNOIS-UHFFFAOYSA-N
SMILES:
ClCC1(CCl)CC1(Br)Br
1
–
1
of
1
results
1
1,1-Dibromo-2,2-bis(chloromethyl)cyclopropane 96.0+%, TCI America™
CAS: 98577-44-7 Molecular Formula: C5H6Br2Cl2 Molecular Weight (g/mol): 296.81 MDL Number: MFCD00101445 InChI Key: RCRVZCIUKQNOIS-UHFFFAOYSA-N PubChem CID: 534192 IUPAC Name: 1,1-dibromo-2,2-bis(chloromethyl)cyclopropane SMILES: ClCC1(CCl)CC1(Br)Br