missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1'-Dibenzoylferrocene 98.0+%, TCI America™
$124.89 - $622.59
Chemical Identifiers
| CAS | 12180-80-2 |
|---|---|
| Molecular Formula | C24H18FeO2 |
| MDL Number | MFCD00045455 |
| Synonym | Bis(benzoylcyclopentadienyl) Iron, Di(benzoylcyclopentadienyl) Iron |
Chemical Identifiers
| 12180-80-2 | |
| C24H18FeO2 | |
| MFCD00045455 | |
| Bis(benzoylcyclopentadienyl) Iron, Di(benzoylcyclopentadienyl) Iron |
Specifications
| 12180-80-2 | |
| C24H18FeO2 | |
| 5 g | |
| 394.25 | |
| Crystalline Powder |
| 103°C | |
| MFCD00045455 | |
| Bis(benzoylcyclopentadienyl) Iron, Di(benzoylcyclopentadienyl) Iron | |
| ≥98.0% (GC) | |
| 1,1′-Dibenzoylferrocene |
Safety and Handling
TSCA : Yes