accessibility menu, dialog, popup

UserName

OFeO

CAS RN 12180-80-2

IUPAC Name: bis(2-benzoylcyclopenta-2,4-dien-1-yl)iron
Synonyms: bis(2-benzoylcyclopenta-2,4-dien-1-yl)iron
Molecular Weight (g/mol): 394.25
Molecular Formula: C24H18FeO2
InChi Key: HAAYOEOQZIDZLZ-UHFFFAOYNA-N
SMILES: O=C(C1=CC=CC1[Fe]C1C=CC=C1C(=O)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results