missing translation for 'onlineSavingsMsg'
Learn More

1,1'-(Decane-1,10-diyl)bis[4-aza-1-azoniabicyclo[2.2.2]octane] Dibromide 95.0+%, TCI America™

Catalog No. d32691g
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D32691G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D32691G Supplier TCI America Supplier No. D32691G
Only null left

Chemical Identifiers

CAS 94630-53-2
Molecular Formula C22H44Br2N4
Molecular Weight (g/mol) 524.43
InChI Key YTGWJQLFKYWCCF-UHFFFAOYSA-L
Synonym 1,10-Bis[4-aza-1-azoniabicyclo[2.2.2]octan-1-yl]decane Dibromide
PubChem CID 44629777
IUPAC Name 4-[10-(1-aza-4-azoniabicyclo[2.2.2]octan-4-yl)decyl]-1-aza-4-azoniabicyclo[2.2.2]octane;dibromide
SMILES C1C[N+]2(CCN1CC2)CCCCCCCCCC[N+]34CCN(CC3)CC4.[Br-].[Br-]

Specifications

Color White
Quantity 1 g
Formula Weight 524.43
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,1′-(Decane-1,10-diyl)bis[4-aza-1-azoniabicyclo[2.2.2]octane] Dibromide