accessibility menu, dialog, popup

UserName

CAS RN 94630-53-2

NNNNBrBr

CAS RN 94630-53-2

IUPAC Name: 1-(10-{1,4-diazabicyclo[2.2.2]octan-1-ium-1-yl}decyl)-1,4-diazabicyclo[2.2.2]octan-1-ium dibromide
Synonyms: 1-(10-{1,4-diazabicyclo[2.2.2]octan-1-ium-1-yl}decyl)-1,4-diazabicyclo[2.2.2]octan-1-ium dibromide
Molecular Weight (g/mol): 524.43
Molecular Formula: C22H44Br2N4
InChi Key: YTGWJQLFKYWCCF-UHFFFAOYSA-L
SMILES: [Br-].[Br-].C(CCCCC[N+]12CCN(CC1)CC2)CCCC[N+]12CCN(CC1)CC2
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results