missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1-Cyclopropanedicarboxylic Acid 98.0+%, TCI America™
$67.11 - $192.00
Chemical Identifiers
| CAS | 598-10-7 |
|---|---|
| Molecular Formula | C5H4O4 |
| Molecular Weight (g/mol) | 128.08 |
| MDL Number | MFCD00013727 |
| InChI Key | FDKLLWKMYAMLIF-UHFFFAOYSA-L |
| Synonym | 1,1-cyclopropanedicarboxylic acid, 1,1-cyclopropanedicarboxylate, cyclopropane-1,1-dicarboxylicacid, cyclopropanedicarboxylic acid, vinaconic acid, cyclopropane, 3, vinyl-malonic acid, 9ty, ethylenemalonic acid, acmc-209mfz |
| PubChem CID | 69017 |
| IUPAC Name | cyclopropane-1,1-dicarboxylate |
| SMILES | [O-]C(=O)C1(CC1)C([O-])=O |
Chemical Identifiers
| 598-10-7 | |
| 128.08 | |
| FDKLLWKMYAMLIF-UHFFFAOYSA-L | |
| 69017 | |
| [O-]C(=O)C1(CC1)C([O-])=O |
| C5H4O4 | |
| MFCD00013727 | |
| 1,1-cyclopropanedicarboxylic acid, 1,1-cyclopropanedicarboxylate, cyclopropane-1,1-dicarboxylicacid, cyclopropanedicarboxylic acid, vinaconic acid, cyclopropane, 3, vinyl-malonic acid, 9ty, ethylenemalonic acid, acmc-209mfz | |
| cyclopropane-1,1-dicarboxylate |
Specifications
| 598-10-7 | |
| White | |
| MFCD00013727 | |
| 3261 | |
| FDKLLWKMYAMLIF-UHFFFAOYSA-L | |
| cyclopropane-1,1-dicarboxylate | |
| 69017 | |
| ≥98.0% (GC,T) | |
| 1,1-Cyclopropanedicarboxylic Acid |
| 137°C | |
| C5H4O4 | |
| 1 g | |
| 1,1-cyclopropanedicarboxylic acid, 1,1-cyclopropanedicarboxylate, cyclopropane-1,1-dicarboxylicacid, cyclopropanedicarboxylic acid, vinaconic acid, cyclopropane, 3, vinyl-malonic acid, 9ty, ethylenemalonic acid, acmc-209mfz | |
| [O-]C(=O)C1(CC1)C([O-])=O | |
| 128.08 | |
| 130.10 | |
| Crystalline Powder |
Safety and Handling
TSCA : No