accessibility menu, dialog, popup

UserName

CAS RN 598-10-7

OOOO

CAS RN 598-10-7

IUPAC Name: cyclopropane-1,1-dicarboxylate
Synonyms: cyclopropane-1,1-dicarboxylate
Molecular Weight (g/mol): 128.08
Molecular Formula: C5H4O4
InChi Key: FDKLLWKMYAMLIF-UHFFFAOYSA-L
SMILES: [O-]C(=O)C1(CC1)C([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)