missing translation for 'onlineSavingsMsg'
Learn More

1,1-Cyclopropanedicarboxylic acid monomethyl ester, 97%

Catalog No. AAH2582803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2582803 1 g
AAH2582806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2582803 Supplier Thermo Scientific Chemicals Supplier No. H2582803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 113020-21-6 | C6H8O4 | 144.126 g/mol

1,1-Cyclopropanedicarboxylic acid monomethyl ester is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1-Cyclopropanedicarboxylic acid monomethyl ester is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 113020-21-6
Molecular Formula C6H8O4
Molecular Weight (g/mol) 144.126
MDL Number MFCD06800496
InChI Key IZYOHLOUZVEIOS-UHFFFAOYSA-N
Synonym 1-methoxycarbonyl cyclopropanecarboxylic acid, 1,1-cyclopropanedicarboxylic acid monomethyl ester, 1-methoxycarbonyl cyclopropane-1-carboxylic acid, cyclopropane-1,1-dicarboxylic acid, methyl ester, 1-methoxycarbonylcyclopropanecarboxylic acid, 1,1-cyclopropanedicarboxylic acid-1-methyl ester, 1-methoxycarbonyl cyclopropanecarboxylicacid, acmc-1c7y7, methyl cyclopropane-1,1-dicarboxylate, 1-methoxycarbonylcyclopropane carboxylic acid
PubChem CID 13861823
IUPAC Name 1-methoxycarbonylcyclopropane-1-carboxylic acid
SMILES COC(=O)C1(CC1)C(=O)O

Specifications

Melting Point 50°C to 57°C
Boiling Point 85°C to 87°C (0.2 mmHg)
Flash Point >110°C (230°F)
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 144.13
Percent Purity 97%
Chemical Name or Material 1,1-Cyclopropanedicarboxylic acid monomethyl ester
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only