Learn More
1-(1-Cyclohexen-1-yl)piperidine, 97%
CAS: 2981-10-4 | C8H22N2O2 | 178.28 g/mol
$92.85 - $270.10
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL0033506
|
Thermo Scientific Chemicals
L0033506 |
5 g |
Each for $92.85
|
|
|||||
|
AAL0033514
|
Thermo Scientific Chemicals
L0033514 |
25 g |
Each for $270.10
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 2981-10-4 | |
| MFCD00014643 |
Specifications
| 2981-10-4 | |
| C8H22N2O2 | |
| MFCD00014643 | |
| 113990 | |
| POTQBGGWSWSMCX-UHFFFAOYSA-P | |
| 3-[2-(3-aminopropoxy)ethoxy]propan-1-amine | |
| 76342 | |
| 97% |
| 120°C to 122°C (16 mmHg) | |
| 1.514 | |
| 5 g | |
| di 3-aminopropoxy ethane, 1-propanamine, 3,3'-1,2-ethanediylbis oxy bis, 1,2-bis 3-aminopropoxy ethane, 3,3'-ethane-1,2-diylbis oxy bis propan-1-amine, 3,3'-ethylenedioxy bispropylamine, 3,3'-ethylenedioxybis propylamine, propylamine, 3,3'-ethylenedioxy bis, ethylene glycol bis 3-aminopropyl ether, 1,10-diamino-4,7-dioxadecane, 3-2-3-aminopropoxy ethoxy propan-1-amine | |
| C(CN)COCCOCCCN | |
| 178.28 | |
| 165.28 | |
| 1-(1-Cyclohexen-1-yl)piperidine |
Safety and Handling
GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
DOTInformation : Hazard Class: 8; Packaging Group: III
RTECSNumber : TM6510000
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only