missing translation for 'onlineSavingsMsg'
Learn More

1-(1-Cyclohexen-1-yl)piperidine, 97%

Catalog No. AAL0033506
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0033506 5 g
AAL0033514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0033506 Supplier Thermo Scientific Chemicals Supplier No. L0033506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2981-10-4 | C8H22N2O2 | 178.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2981-10-4
MDL Number MFCD00014643

Specifications

Boiling Point 120°C to 122°C (16 mmHg)
Molecular Formula C8H22N2O2
Refractive Index 1.514
Quantity 5 g
Beilstein 113990
Synonym di 3-aminopropoxy ethane, 1-propanamine, 3,3'-1,2-ethanediylbis oxy bis, 1,2-bis 3-aminopropoxy ethane, 3,3'-ethane-1,2-diylbis oxy bis propan-1-amine, 3,3'-ethylenedioxy bispropylamine, 3,3'-ethylenedioxybis propylamine, propylamine, 3,3'-ethylenedioxy bis, ethylene glycol bis 3-aminopropyl ether, 1,10-diamino-4,7-dioxadecane, 3-2-3-aminopropoxy ethoxy propan-1-amine
InChI Key POTQBGGWSWSMCX-UHFFFAOYSA-P
SMILES C(CN)COCCOCCCN
IUPAC Name 3-[2-(3-aminopropoxy)ethoxy]propan-1-amine
Molecular Weight (g/mol) 178.28
PubChem CID 76342
Formula Weight 165.28
Percent Purity 97%
Chemical Name or Material 1-(1-Cyclohexen-1-yl)piperidine
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
RTECSNumber TM6510000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only