missing translation for 'onlineSavingsMsg'
Learn More

1,1-Cyclohexanediacetic acid, 98%

Catalog No. AAB2081609
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2081609 10 g
AAB2081618 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2081609 Supplier Thermo Scientific Chemicals Supplier No. B2081609
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4355-11-7 | C10H16O4 | 200.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4355-11-7
Molecular Formula C10H16O4
Molecular Weight (g/mol) 200.23
MDL Number MFCD00001519
InChI Key YQPCHPBGAALCRT-UHFFFAOYSA-N
Synonym 1,1-cyclohexanediacetic acid, 1,1-cyclohexanediaceticacid, cyclohexanediacetic acid, 2-1-carboxymethyl cyclohexyl acetic acid, 1,1-cyclohexane diacetic acid, 1-carboxymethyl cyclohexyl acetic acid, 2,2'-cyclohexane-1,1-diyl diacetic acid, 2-carboxymethyl cyclohexyl acetic acid, gabapentin impurity 2, pubchem13645
PubChem CID 78061
IUPAC Name 2-[1-(carboxymethyl)cyclohexyl]acetic acid
SMILES C1CCC(CC1)(CC(=O)O)CC(=O)O

Specifications

Melting Point 178°C to 182°C
Quantity 10 g
Formula Weight 200.23
Percent Purity 98%
Chemical Name or Material 1,1-Cyclohexanediacetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 224-427-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only