missing translation for 'onlineSavingsMsg'
Learn More

1,1-Bis(4-aminophenyl)cyclohexane 98.0+%, TCI America™

Catalog No. B20765G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B20765G 5 g
B207625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B20765G Supplier TCI America Supplier No. B20765G
Only null left

Chemical Identifiers

CAS 3282-99-3
Molecular Formula C18H22N2
Molecular Weight (g/mol) 266.39
MDL Number MFCD00035700
InChI Key DJCVCYUEICJZBD-UHFFFAOYSA-N
PubChem CID 768325
IUPAC Name 4-[4-(4-aminophenyl)cyclohexyl]aniline
SMILES NC1=CC=C(C=C1)C1CCC(CC1)C1=CC=C(N)C=C1

Specifications

Melting Point 110°C
Quantity 5 g
Synonym 4,4′C-Cyclohexylidenedianiline
Formula Weight 266.39
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,1-Bis(4-aminophenyl)cyclohexane
EINECSNumber (4)-0045
TSCA No