missing translation for 'onlineSavingsMsg'
Learn More

1,1'-Bis(2,4-dinitrophenyl)-4,4'-bipyridinium Dichloride 97.0+%, TCI America™

Catalog No. p-7119089
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
B15185G 5 g
B151825G 25 g
2 options

Catalog No. B15185G

Supplier: TCI America B15185G

Only null left

Chemical Identifiers

CAS 41168-79-0
Molecular Formula C22H14Cl2N6O8
Molecular Weight (g/mol) 561.288
MDL Number MFCD00142352
InChI Key HTRYWXJIGDXDLF-UHFFFAOYSA-L
PubChem CID 14228160
IUPAC Name 1-(2,4-dinitrophenyl)-4-[1-(2,4-dinitrophenyl)pyridin-1-ium-4-yl]pyridin-1-ium;dichloride
SMILES C1=CC(=C(C=C1[N+](=O)[O-])[N+](=O)[O-])[N+]2=CC=C(C=C2)C3=CC=[N+](C=C3)C4=C(C=C(C=C4)[N+](=O)[O-])[N+](=O)[O-].[Cl-].[Cl-]

Specifications

Color Yellow
Quantity 5 g
UN Number 1325
Formula Weight 561.29
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1,1′-Bis(2,4-dinitrophenyl)-4,4′-bipyridinium Dichloride