accessibility menu, dialog, popup

UserName

CAS RN 41168-79-0

NOONOONNNOONOOClCl

CAS RN 41168-79-0

IUPAC Name: 1,1'-bis(2,4-dinitrophenyl)-1λ⁵,1'λ⁵-[4,4'-bipyridine]-1,1'-bis(ylium) dichloride
Synonyms: 1,1'-bis(2,4-dinitrophenyl)-1λ⁵,1'λ⁵-[4,4'-bipyridine]-1,1'-bis(ylium) dichloride
Molecular Weight (g/mol): 561.29
Molecular Formula: C22H14Cl2N6O8
InChi Key: HTRYWXJIGDXDLF-UHFFFAOYSA-L
SMILES: [Cl-].[Cl-].[O-][N+](=O)C1=CC(=C(C=C1)[N+]1=CC=C(C=C1)C1=CC=[N+](C=C1)C1=C(C=C(C=C1)[N+]([O-])=O)[N+]([O-])=O)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results