missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1'-Bis(2,4-dinitrophenyl)-4,4'-bipyridinium Dichloride 97.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Quantity | 25 g |
| UN Number | 1325 |
| Formula Weight | 561.29 |
| Percent Purity | ≥97.0% (HPLC) |
| Grade | Reagent |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 1,1′-Bis(2,4-dinitrophenyl)-4,4′-bipyridinium Dichloride |
Safety and Handling
| TSCA | No |