missing translation for 'onlineSavingsMsg'
Learn More

1,1'-Biadamantane, Thermo Scientific Chemicals

Catalog No. AC465932500
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC465932500 250 mg
AC465930010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC465932500 Supplier Thermo Scientific Chemicals Supplier No. 465932500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3732-31-8
Molecular Formula C20H30
Molecular Weight (g/mol) 270.46
MDL Number MFCD01910632
InChI Key MPXKIFWZOQVOLN-UHFFFAOYSA-N
Synonym 1,1 '-Diadamantane; Bisadamantane
IUPAC Name 1,1'-biadamantane
SMILES C1C2CC3CC1CC(C2)(C3)C12CC3CC(CC(C3)C1)C2

Specifications

Alpha Vector BIADAMANTANE
Melting Point 285°C to 295°C
Color White
Infrared Spectrum Conforms
Packaging glass bottle
Quantity 250 mg
Formula Weight 270.46
Physical Form Powder
Chemical Name or Material 1,1 '-Biadamantane
ShelfLife 5 years
storageNote1 Refrigerator +4°C

RUO – Research Use Only