accessibility menu, dialog, popup

UserName

CAS RN 3732-31-8

IUPAC Name: 1,1'-biadamantane
Synonyms: 1,1'-biadamantane
Molecular Weight (g/mol): 270.46
Molecular Formula: C20H30
InChi Key: MPXKIFWZOQVOLN-UHFFFAOYSA-N
SMILES: C1C2CC3CC1CC(C2)(C3)C12CC3CC(CC(C3)C1)C2
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)